Learn More
Sino Biological Recombinant Human Carbonic Anhydrase III / CAIII Protein (His Tag)
Supplier:  Sino Biological 10503H08E100
Encompass_Preferred
A DNA sequence encoding the human CA3 (NP_005172.1) (Met 1-Lys 260) was expressed, with a polyhistide tag at the C-terminus.
Catalog No. 50-161-5082