Learn More
Sino Biological Recombinant Human PDGF beta receptor/PDGFRB Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10514H08H100
Encompass_Preferred
A DNA sequence encoding the human PDGFRβ (NP_002600.1) extracellular domain (Met 1-Lys 531) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-5100