Learn More
Sino Biological Recombinant Human IL-3R Beta/IL3RB Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10516H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Trp 443) of human CSF2RB (NP_000386.1) expressed, fused with a polyhistidine-tag at the C-terminus.
Catalog No. 50-161-5105