Learn More
Sino Biological Recombinant Human ADAM15 Protein (His Tag)
Supplier:  Sino Biological 10517H08H20
Encompass_Preferred
A DNA sequence encoding the human ADAM15 (NP_997074.1) (Met 1-Thr 696) precursor with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5108