Learn More
Sino Biological Recombinant Human LBP Protein (His Tag)
Supplier:  Sino Biological 10526H08H50
Encompass_Preferred
A DNA sequence encoding the human LBP (NP_004130.2) extracellular domain (Met 1-Val 481) was fused with the a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5120