Learn More
Sino Biological Recombinant Human RELT/TNFRSF19L Protein (His & hFc Tag)
Supplier:  Sino Biological 10530H03H100
Encompass_Preferred
A DNA sequence encoding the human RELT (NP_116260.2) extracellular domain (Met 1-Ala 160) with 127R/G & 129R/G mutation was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5127