Learn More
Sino Biological Recombinant Human RELT/TNFRSF19L Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10530H08H100
Encompass_Preferred
A DNA sequence encoding the human RELT (NP_116260.2) extracellular domain (Met 1-Ala 160) was fused with a polyhistidine tag at the C-terminus, with 127R/G & 129R/G mutations.
Catalog No. 50-161-5129