Learn More
Sino Biological Recombinant Human Chk1 Protein (GST Tag)
Supplier:  Sino Biological 10539H09B50
Encompass_Preferred
A DNA sequence encoding the human CHK1 (AAM78553.1) (Met1-Thr476) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5144