Learn More
Sino Biological Recombinant Human ACP5 Protein (His Tag)
Supplier:  Sino Biological 10550H08H20
Encompass_Preferred
A DNA sequence encoding the human ACP5 (NP_001602.1) precursor (Met 1-Pro 320) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5156