Learn More
Sino Biological Recombinant Human BMPR2 Protein (His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 10551H03H200
Encompass_Preferred
A DNA sequence encoding the human BMPR-II (NP_001195.2) extracellular domain (Met 1-Ile 151) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5158