Learn More
Sino Biological Recombinant Human PDGF alpha receptor/PDGFRA Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10556H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 524) of human PDGFRα (NP_006197.1) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5167