Learn More
Sino Biological Recombinant Human DC-SIGNR/CD299 Protein (ECD)
Supplier:  Sino Biological 10559HNCH50
Encompass_Preferred
A DNA sequence encoding the human CLEC4M (NP_055072.3) (Ser78-Glu399) was expressed.
Catalog No. 50-161-5180