Learn More
Sino Biological Recombinant Human Bcl2A1 Protein (His Tag)
Supplier:  Sino Biological 10562H07E200
Encompass_Preferred
A DNA sequence encoding the human BCL2A1 (NP_004040 ) (Met1-Ser152) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-5186