Learn More
Sino Biological Recombinant Human ANGPTL4 Protein (hFc Tag)
Supplier:  Sino Biological 10563H01H100
Encompass_Preferred
A DNA sequence encoding the human ANGPTL4 (Q9BY76) (Pro166-Ser406) was expressed,with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-5188