Learn More
Sino Biological Recombinant Human ANGPTL4 Protein (His Tag)
Supplier:  Sino Biological 10563H07H100
Encompass_Preferred
A DNA sequence encoding the human ANGPTL4 (Q9BY76) (Pro166-Ser406) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-161-5190