Learn More
Sino Biological Recombinant Human DNAM-1/CD226 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10565H08H100
Encompass_Preferred
A DNA sequence encoding the human DNAM1 (NP_006557.2) extracellular domain (Met1-Asn247) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5198