Learn More
Sino Biological Recombinant Human Beta ig-h3/BIGH3/TGFBI Protein (His Tag)
Supplier:  Sino Biological 10569H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human beta IG-H3 (NP_000349.1) precursor (Met 1-His 683) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5209