Learn More
Sino Biological Recombinant Human TC-PTP/PTPN2 Protein
Supplier:  Sino Biological 10570HNCB100
Encompass_Preferred
A DNA sequence encoding the human PTPN2 ( P17706-1) (Met 1-Asn 314) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-5213