Learn More
Sino Biological Recombinant Human PDGF-B Protein (His Tag)
Supplier:  Sino Biological 10572H07Y50
Encompass_Preferred
A DNA sequence encoding the human PDGFB (NP_002599.1) (Ser82-Thr190) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-5217