Learn More
Sino Biological Recombinant Human FGF-10 Protein, HPLC-verified
Supplier:  Sino Biological 10573HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human FGF10 (NP_004456.1) (Gln38-Ser208) was expressed with an N-terminal Met.
Catalog No. 50-161-5218