Learn More
Sino Biological Recombinant Human NKG2D Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10575H01S100
Encompass_Preferred
A DNA sequence encoding the human NKG2D (NP_031386.2) (Phe78-Val216) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-5221