Learn More
Sino Biological Recombinant Human NKG2D Protein (aa 78-216, His Tag), HPLC-verified
Supplier:  Sino Biological 10575H07B100
Encompass_Preferred
A DNA sequence encoding the human NKG2D (P26718-1) (Phe78-Val216) was fused with a polyhistide tag at the N-terminus.
Catalog No. 50-161-5223