Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Phytohemagglutinin, M form (PHA-M)

Catalog No. 10576015
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-576-015 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-576-015 Supplier Gibco™ Supplier No. 10576015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This extract is capable of inducing blastogenesis in vitro in various mammalian mononuclear cells when used at a concentration of 1 to 2% v/v.

Gibco™ Phytohemagglutinin, M form (PHA-M) is a solution made from a crude extract of the red kidney bean Phaseolus vulgaris. This extract is capable of inducing blastogenesis in vitro in various mammalian mononuclear cells when used at a concentration of 1–2% v/v. Each lot of Gibco™ Phytohemagglutinin is performance tested at 1.5% using peripheral blood lymphocytes from a normal adult donor cultured for 72 hours in Gibco™ RPMI 1640 Medium supplemented with Fetal Bovine Serum (FBS), heparin, antibiotics, and L-glutamine.

cGMP Manufacturing and Quality System

Gibco™ Phytohemagglutinin is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Baculovirus Production and Titering, Cell Culture, Insect Cell Culture, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5°C to -20°C. Protect from light.
Shipping conditions: Dry ice
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Form Liquid
Product Type Phytohemagglutinin
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Quantity 10 mL
Shipping Condition Dry Ice
What is the concentration of the PHA-M solution?

The PHA is a crude extract of red kidney beans. No protein concentration assay is performed. We recommend using the product at 1-2% by volume, or you may choose to optimize the dose by titration.

For how long is Phytohemagglutinin, M form (PHA-M) stable one thawed. Can I freeze it again?

PHA-M is shipped on dry ice and recommended storage is at -5 degrees C to -20 degrees C.

Post-thawing, the product should be used within 10-15 days. We do not recommend freezing it back.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.