Learn More
Sino Biological Recombinant Human IL-21 Protein, HPLC-verified
Supplier:  Sino Biological 10584HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human IL-21 (Q9HBE4-1) (Gln30-Ser162) was expressed with an initial Met.
Catalog No. 50-161-5238