Learn More
Sino Biological Recombinant Human GRO Beta/CXCL2 Protein
Supplier:  Sino Biological 10586HNCE10
Encompass_Preferred
A DNA sequence encoding the human CXCL2 (NP_002080.1) (Ala35-Asn107) was expressed.
Catalog No. 50-161-5244