Learn More
Sino Biological Recombinant Human SOST/Sclerostin Protein (His Tag)
Supplier:  Sino Biological 10593H07H50
Encompass_Preferred
A DNA sequence encoding the mature form of human SOST (NP_079513.1) (Gln24-Tyr213) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-5250