Learn More
Sino Biological Human PCSK9 / NARC1 (D374Y, V474I, G670E) Protein (His Tag)
Supplier:  Sino Biological 10594H08H150
Encompass_Preferred
A DNA sequence encoding the human PCSK9 (NP_777596.2, with mutation Asp 374 Tyr, natural mutation Val 474 Ile and Gly 670 Glu) (Met 1-Gln 692) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5253