Learn More
Sino Biological Recombinant Rat ALK-1 Protein (His & hFc Tag)
Supplier:  Sino Biological 10596R03H200
Encompass_Preferred
A synthetic DNA sequence encoding the rat ACVRL1 (NP_071886.1) extracellular domain (Met 1-Ala 118) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-5257