Learn More
Hanna Pharmaceutical Supply Co., Inc. CONTINU-FLO Solution Set 10dr/mL 104""""
Catalog No. NC1842692
Encompass_Preferred
Due to recent natural disasters, production of sterile water has been negatively impacted.  Please expect long lead times on these itemsCONTINU-FLO Solution Set with Non-Vented Spike, 0.2 Micron Filter, 2 CLEARLINK Luer Activated Valve, Backcheck Valve and Male Luer Lock Adapter with Retractable Collar. Approximately 10 drops/mL. Approximate Length 104" (2.6 m). Contains DEHP. Fluid path is sterile, nonpyrogenic. 48/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC1842692 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BAX2C8571
Due to recent natural disasters, production of sterile water has been negatively impacted.  Please expect long lead times on these itemsCONTINU-FLO Solution Set with Non-Vented Spike, 0.2 Micron Filter, 2 CLEARLINK Luer Activated Valve, Backcheck Valve and Male Luer Lock Adapter with Retractable Collar. Approximately 10 drops/mL. Approximate Length 104" (2.6 m). Contains DEHP. Fluid path is sterile, nonpyrogenic. 48/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.