Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gasco Oxygen | 7782-44-7 | MFCD00011434 | 105L

Catalog No. NC1716866
Encompass_Preferred

Oxygen | 70ppm | MW: 31.998 | 7782-44-7 | MFCD00011434 | 105L

Catalog No. NC1716866 Supplier Gasco Supplier No. 105L15970
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxygen | 70ppm | MW: 31.998 | 7782-44-7 | MFCD00011434 | 105L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.