Learn More
Sino Biological Recombinant Human Complement C5 Protein (C5a), HPLC-verified
Supplier:  Sino Biological 10604HNAE50
Encompass_Preferred
A DNA sequence encoding the active form of human C5a (NP_001726.2) (Leu679-Arg751) was expressed.
Catalog No. 50-161-5272