Learn More
Sino Biological Recombinant Human EGF Protein (ECD, hFc Tag)
Supplier:  Sino Biological 10605H01H100
Encompass_Preferred
A DNA sequence encoding the mature soluble form of human EGF (NP_001954.2) (Asn 971-Arg 1023) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-5274