Learn More
Sino Biological Recombinant Human IL-36 alpha/IL-1F6 Protein (aa 6-158)
Supplier:  Sino Biological 10607HNCE120
Encompass_Preferred
A DNA sequence encoding the human IL1F6 (Q9UHA7) (Ala6-Phe158) was expressed and purified, with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-5291