Learn More
Sino Biological Recombinant Human FGFR1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10616H08H100
Encompass_Preferred
A DNA sequence encoding the human FGFR1 (NP_075594.1) extracellular domain (Met 1-Glu 285) was expressed with C-terminal polyhistidine tag.
Catalog No. 50-161-5296