Learn More
Sino Biological Recombinant Human HtrA2/Omi Protein (His Tag)
Supplier:  Sino Biological 10619H08E100
Encompass_Preferred
A DNA sequence encoding the mature form of human HTRA2 (O43464-1) (Ala 134-Glu 458) was expressed, with a polyhistide tag at the C-terminus.
Catalog No. 50-161-5304