Learn More
Sino Biological Recombinant Human BCA-1/CXCL13 Protein
Supplier:  Sino Biological 10621HNAE20
Encompass_Preferred
A DNA sequence encoding the human CXCL13 (NP_006410.1) (Val23-Arg94) was expressed with an initial Met.
Catalog No. 50-161-5313