Learn More
Sino Biological Recombinant Human CDK2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10624H08B50
Encompass_Preferred
A DNA sequence encoding the human cyclin-dependent kinase 2 isoform 1 (NP_001789.2) (Met 1-Leu 298) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-5317