Learn More
Sino Biological Recombinant Human LIFR Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10628H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human LIFR (NP_001121143.1) (Met 1-Ser 833) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5320