Learn More
Sino Biological Recombinant Human CK2 alpha/CSNK2A1 Protein (GST Tag)
Supplier:  Sino Biological 10630H09B50
Encompass_Preferred
A DNA sequence encoding the human CSNK2A1 isoform 1 (NP_808227.1) (Met 1-Gln 391) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5323