Learn More
Sino Biological Recombinant Human Fractalkine/CX3CL1 Protein (His Tag)
Supplier:  Sino Biological 10636H08H50
Encompass_Preferred
A DNA sequence encoding the human CX3CL1 (NP_002987.1) extracellular domain (Met 1-Arg 339) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5327