Learn More
Sino Biological Recombinant Human SPOCK1 Protein (His Tag)
Supplier:  Sino Biological 10644H08B50
Encompass_Preferred
A DNA sequence encoding the human SPOCK1 (Q08629) ( Met 1-Trp439) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5331