Learn More
Sino Biological Recombinant Human CDC37 Protein (GST Tag)
Supplier:  Sino Biological 10647H09B50
Encompass_Preferred
A DNA sequence encoding the full length of human CDC37 (NP_008996.1) (Met 1-Val 378) was fused with the GST tag at the C-terminus.
Catalog No. 50-161-5335