Learn More
Sino Biological Recombinant Human RAF1/c-Raf Protein (His & GST Tag)
Supplier:  Sino Biological 10657H20B50
Encompass_Preferred
A DNA sequence encoding the human RAF1 (NP_002871.1) (Ser306-Phe648) was expressed with a N-terminal GST tagged polyhistidine tag at the N-terminus.
Catalog No. 50-161-5346