Learn More
Sino Biological Recombinant Human Neuregulin-1/NRG1 Protein (hFc Tag)
Supplier:  Sino Biological 10658H01H100
Encompass_Preferred
A DNA sequence encoding the human NRG1 (NP_001153480.1) (Ser177-Gln237) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-5347