Learn More
Sino Biological Recombinant Human Azurocidin/CAP37 Protein (His Tag)
Supplier:  Sino Biological 10660H08H100
Encompass_Preferred
A DNA sequence encoding the human AZU1 (NP_001691.1) (Met 1-Pro 250) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-5352