Learn More
Sino Biological Recombinant Human PKC nu Protein (GST Tag)
Supplier:  Sino Biological 10665H09B50
Encompass_Preferred
A DNA sequence encoding the full length of human PRKD3 (NP_005804.1) (Met 1-Pro 890) was expressed with the GST tag at the N-terminus.
Catalog No. 50-161-5357