Learn More
Sino Biological Recombinant Human Epiregulin Protein (hFc Tag)
Supplier:  Sino Biological 10677H01H100
Encompass_Preferred
A DNA sequence encoding the mature form of human EREG (NP_001423.1) (Val63-Leu108) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-5368