Learn More
Sino Biological Recombinant Human SFRP1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10680H08H50
Encompass_Preferred
A DNA sequence encoding the human sFRP1 (NP_003003.3) (Met 1-Lys 314) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-5378