Learn More
Sino Biological Recombinant Human Brk Protein (GST Tag)
Supplier:  Sino Biological 10682H09B50
Encompass_Preferred
A DNA sequence encoding the full length of human PTK6 (NP_005966.1) (Met 1-Thr 451) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-5380