Learn More
Sino Biological Recombinant Human MAPKAPK3 Protein (GST Tag)
Supplier:  Sino Biological 10683H09B50
Encompass_Preferred
A DNA sequence encoding the full length of human MAPKAPK3 (NP_004626.1) (Met 1-Gln 382) was expressed with the GST tag at the N-terminus.
Catalog No. 50-161-5382